missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RIIB/CD32b Antibody (110), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18323550 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323550 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323550 Supplier Novus Biologicals Supplier No. NBP289364AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Fc gamma RIIB/CD32b Monoclonal specifically detects Fc gamma RIIB/CD32b in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fc gamma RIIB/CD32b
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 110
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Fc gamma RIIB/CD32b extracellular domain (Ala 46-Pro 217; NP_001002274.1).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2213
Test Specificity Has cross-reactivity in ELISA with: Human CD64 Human CD16b No cross-reactivity in ELISA with: Human CD32a Human CD16a Mouse FCGR2B
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.