missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Fc gamma RII/RIII (CD32/CD16) Antibody (J5511), Novus Biologicals™
Mouse Monoclonal Antibody
£162.00 - £247.00
Specifications
| Antigen | Fc gamma RII/RIII (CD32/CD16) |
|---|---|
| Clone | J5511 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067162
|
Novus Biologicals™
NBP3-30971-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066890
|
Novus Biologicals™
NBP3-30971-100ug |
100 μg |
£247.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RII/RIII (CD32/CD16) Monoclonal antibody specifically detects Fc gamma RII/RIII (CD32/CD16) in Human samples. It is validated for Flow CytometrySpecifications
| Fc gamma RII/RIII (CD32/CD16) | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Immunology | |
| Phosphate buffered saline | |
| 2214 | |
| IgG1 | |
| Protein A or G purified |
| J5511 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD16;CD16A;CD16-II;FCG3;FCGR3;FCGR3A;FCGRIII;FcGRIIIA;FCR-10;FCRIII;FCRIIIA;IGFR3;IMD20;Low affinity immunoglobulin gamma Fc region receptor III-A | |
| CD16=Derived from the hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells of BALB/c mice immunized with NK lymphocytes | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title