missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fc gamma RII/CD32 Monoclonal antibody specifically detects Fc gamma RII/CD32 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Functional Assay, Neutralization
Specifications
Specifications
| Antigen | Fc gamma RII/CD32 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Functional Assay, Neutralization |
| Classification | Monoclonal |
| Clone | AT130-5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional, Block/Neutralize |
| Formulation | PBS |
| Gene Alias | CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b |
| Host Species | Rat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?