missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RII/CD32 Mouse anti-Human, Clone: FCGR2A/479, Novus Biologicals™
Mouse Monoclonal Antibody
£215.00
Specifications
| Antigen | Fc gamma RII/CD32 |
|---|---|
| Clone | FCGR2A/479 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Functional |
| Applications | Flow Cytometry, Immunofluorescence, Functional Assay |
Description
Fc gamma RII/CD32 Monoclonal specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional.Specifications
| Fc gamma RII/CD32 | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunofluorescence, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
| Recombinant human Fc gamma RII/CD32 protein | |
| Primary | |
| Store at 4°C. |
| FCGR2A/479 | |
| Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Functional | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 2213 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title