missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RII/CD32 Antibody (AT130-5) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
£216.00 - £467.00
Specifications
| Antigen | Fc gamma RII/CD32 |
|---|---|
| Clone | AT130-5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Functional Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620169
|
Novus Biologicals
NBP3-08997-0.2mg |
0.2 mg |
£467.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662149
|
Novus Biologicals
NBP3-08997-0.025mg |
0.025 mg |
£216.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RII/CD32 Monoclonal antibody specifically detects Fc gamma RII/CD32 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Functional Assay, NeutralizationSpecifications
| Fc gamma RII/CD32 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
| CD32-rat CD4 domain 3/4 fusion protein. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| AT130-5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Functional Assay | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS | |
| 2213 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title