missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RII/CD32 Antibody (AT10) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody

£206.00 - £498.00

Specifications

Antigen Fc gamma RII/CD32
Clone AT10
Concentration 1 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18628407
View Documents
Novus Biologicals
NBP3-08998-0.2mg
0.2 mg
£498.00
0.20mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18686107
View Documents
Novus Biologicals
NBP3-08998-0.025mg
0.025 mg
£206.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fc gamma RII/CD32 Monoclonal antibody specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

Fc gamma RII/CD32
1 mg/mL
Monoclonal
Purified
RUO
Human
CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b
Human CD32.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
AT10
Flow Cytometry
Unconjugated
Rabbit
Immunology
PBS
2213
IgG κ
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.