missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RII/CD32 Antibody (AT10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
£170.00 - £436.00
Specifications
| Antigen | Fc gamma RII/CD32 |
|---|---|
| Clone | AT10 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 20 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 20 ug/ml, Immunohistochemistry-Frozen 1:500-1:1000 |
| Applications | Flow Cytometry, Immunoprecipitation, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18001436
|
Novus Biologicals
NB100-65338-0.025MG |
0.025 mg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18730414
|
Novus Biologicals
NB100-65338 |
0.1 mg |
£436.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RII/CD32 Monoclonal specifically detects Fc gamma RII/CD32 in Human, Porcine, Canine, Rhesus Macaque samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| Fc gamma RII/CD32 | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunoprecipitation, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| P12318 | |
| 2213 | |
| K562 cell line | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| AT10 | |
| Flow Cytometry 20 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 20 ug/ml, Immunohistochemistry-Frozen 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Pig, Primate, Canine | |
| CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
| FCGR2B | |
| IgG1 | |
| Protein A purified | |
| NB100-65338 recognizes the human CD32 antigen, a 40kD glycoprotein that acts as a low affinity receptor for IgG (also known as Fc gamma RII). The antigen mediates several functions including endocytosis, activation of secretion, cytotoxicity and immunomodulation. CD32 is expressed by B cells, monocytes, granulocytes and platelets. Clone AT10 blocks the binding of IgG to Fc gamma RII. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title