missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RI/CD64, Human anti-Human, Clone: H22, Novus Biologicals™
Human Monoclonal Antibody
£217.00 - £463.00
Specifications
| Antigen | Fc gamma RI/CD64 |
|---|---|
| Clone | H22 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, Competitive ELISA |
| Applications | Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18249859
|
Novus Biologicals
NBP2-81097-0.025MG |
0.025 mg |
£217.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18353761
|
Novus Biologicals
NBP2-81097-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Competitive ELISA.Specifications
| Fc gamma RI/CD64 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Human | |
| Human | |
| CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| FCGR1A | |
| IgG1 κ | |
| Protein A purified | |
| This immunotoxin is specific for human Fc gamma RI/CD64, and specifically an epitope outside the Fc binding domain. The binding activity of the VH/VL antibody format was unaffected by fusion of the m22(scFv) coding regions to the truncated ETA coding sequences |
| H22 | |
| Flow Cytometry, Competitive ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 2209 | |
| This recombinant antibody was prepared by cloning the single chain fragment (scFv) derived from the murine anti-human Fc gamma RI/CD64 mAb m22 into the bacterial expression vector pBM1.1, and fusing it to a deletion mutant of Pseudomonas exotoxin A. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title