missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Fc gamma RI/CD64 Antibody (Medarex Patent Anti-Fc-Gamma-R1), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28132-100ug
This item is not returnable.
View return policy
Description
Fc gamma RI/CD64 Monoclonal antibody specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Fc gamma RI/CD64 | |
| Monoclonal | |
| Unconjugated | |
| P12314 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Medarex Patent Anti-Fc-Gamma-R1 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| Fc gamma R1 | |
| 100 μg | |
| CD Markers, Immunology | |
| 2209 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction