missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RI/CD64 Antibody (H22), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18432117 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432117 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432117 Supplier Novus Biologicals Supplier No. NBP281098MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Fc gamma RI/CD64 Monoclonal antibody specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fc gamma RI/CD64
Applications Flow Cytometry
Classification Monoclonal
Clone H22
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I
Host Species Rabbit
Immunogen This recombinant antibody was prepared by cloning the single chain fragment (scFv) derived from the murine anti-human Fc gamma RI/CD64 mAb m22 into the bacterial expression vector pBM1.1, and fusing it to a deletion mutant of Pseudomonas exotoxin A.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2209
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.