missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Competitive ELISA.
Specifications
Specifications
| Antigen | Fc gamma RI/CD64 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Clone | H22 |
| Conjugate | DyLight 488 |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Competitive ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I |
| Gene Symbols | FCGR1A |
| Host Species | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?