missing translation for 'onlineSavingsMsg'
Learn More

Fc epsilon RI Antibody (IE7), PerCP, Novus Biologicals™

Product Code. 18345553 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345553 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345553 Supplier Novus Biologicals Supplier No. NBP312051PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fc epsilon RI Monoclonal antibody specifically detects Fc epsilon RI in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fc epsilon RI
Applications Flow Cytometry
Classification Monoclonal
Clone IE7
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS
Gene Alias alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide
Host Species Mouse
Immunogen Recombinant human protein, N-terminus.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Asthma, Cancer, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2205
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.