missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc epsilon RI Antibody (AER-37) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£460.00
Specifications
| Antigen | Fc epsilon RI |
|---|---|
| Clone | AER-37 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry-Paraffin |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
Description
Fc epsilon RI Monoclonal specifically detects Fc epsilon RI in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Fc epsilon RI | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide | |
| FCER1A | |
| IgG2b κ | |
| Affinity Purified |
| AER-37 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Mouse | |
| Apoptosis, Asthma, Cancer, Immunology, Signal Transduction | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 2205 | |
| The immunogen for this antibody was FceRI alpha. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title