missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fc epsilon RI Monoclonal antibody specifically detects Fc epsilon RI in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antigen | Fc epsilon RI |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 9E1 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide |
| Host Species | Mouse |
| Immunogen | Made to Human Fc EPSILON R1 ALPHA |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?