missing translation for 'onlineSavingsMsg'
Learn More

Fc epsilon RI alpha Antibody [DyLight 488], Novus Biologicals™

Product Code. 30733365 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733365 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733365 Supplier Novus Biologicals Supplier No. AF6678K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Fc epsilon RI alpha Polyclonal antibody specifically detects Fc epsilon RI alpha in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fc epsilon RI alpha
Applications Flow Cytometry, CyTOF
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Fc epsilon RI alpha-chain, Fc fragment of IgE receptor Ia, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc IgE receptor, alpha polypeptide, FCE1A, Fc-epsilon RI-alpha, FcepsilonRI alpha chain, FcepsilonRIalpha, FCERI, FceRIa, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, igE Fc receptor subunit alpha, immunoglobulin E receptor, high-affinity, of mast cells, alpha polypeptide
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fc epsilon RI alpha , Val26-Gln205, Accession # NP_001992
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2205
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.