Learn More
BD Biosciences™ FC Beads V500-C (RUO)
Fluorescent beads that enable the software to determine spillover values (SOVs) for fluorescence compensation and calculate a fluorescence compensation matrix during setup of BD™ flow cytometers.
Brand: BD Biosciences™ 661624
Additional Details : Weight : 0.50000kg
Description
- Fluorescent beads that enable the software to determine spillover values (SOVs) for fluorescence compensation and calculate a fluorescence compensation matrix during setup of BD™ flow cytometers.
- The combination of BD™ CS&T beads and BD FC beads accurately sets the voltage and compensation for each of the channels of the instrument.
- BD FC beads can also be used to standardize multiple instruments.
Specifications
Store tubes at 2°C–8°C in the foil pouch. The tubes should not be frozen. Protect the tubes from exposure to light and humidity. | |
The beads and dilution buffer are stable until the expiration date shown on the pouch and bottle labels when stored as directed. Do not use after the expiration date. | |
Fluorescent Polystyrene Beads |
V500-C | |
5 Tests | |
3 μm |
Safety and Handling
- BD FC Beads V500-C
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 2-methylisothiazol-3(2H)-one