missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ FC Beads PE-Cy™7 (RUO)
Click to view available options
Quantity:
5 Tests
Unit Size:
5 tests
Description
- Fluorescent beads that enable the software to determine spillover values (SOVs) for fluorescence compensation and calculate a fluorescence compensation matrix during setup of BD™ flow cytometers.
- The combination of BD™ CS&T beads and BD FC beads accurately sets the voltage and compensation for each of the channels of the instrument.
- BD FC beads can also be used to standardize multiple instruments.
Specifications
Specifications
| Content And Storage | Store tubes at 2°C–8°C in the foil pouch. The tubes should not be frozen. Protect the tubes from exposure to light and humidity. |
| Format | PE-Cy™7 |
| No. of Tests | 5 Tests |
| Particle Size | 3 μm |
| Quantity | 5 Tests |
| Stability | The beads and dilution buffer are stable until the expiration date shown on the pouch and bottle labels when stored as directed. Do not use after the expiration date. |
| Product Type | Fluorescent Polystyrene Beads |
Safety and Handling
Product Identifier
- BD FC Beads PE-Cy7
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 2-methylisothiazol-3(2H)-one
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction