missing translation for 'onlineSavingsMsg'
Learn More

FC Beads Dilution Buffer (RUO), BD Biosciences™

Product Code. 15917842
Change view
Click to view available options
Quantity:
35 mL
Unit Size:
35mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15917842 35 mL 35mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15917842 Supplier BD Biosciences Supplier No. 661614
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Suitable for proper and safe dilution and rehydration of FC beads for use.

  • Safely and properly dilutes and rehydrates beads.
  • Contains phosphate buffered saline (PBS) with protein stabilizers and 0.1% sodium azide.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate buffered saline (PBS) with protein stabilizers and 0.1% sodium azide
Quantity 35 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.