Learn More
BD Biosciences™ FC Bead Violet Research Kit (RUO)
Product Code. 15349421
Click to view available options
:
To set compensation for the BD Horizon™ V450 dye on BD flow cytometers with a violet laser.
Product Code. 15349421
Brand: BD Biosciences™ 650627
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used to determine spillover values for reference settings.
  • Designed for use on BD™ flow cytometers running BD FACSuite™ software.
  • FC beads enable the software to automatically determine the spillover values for reference settings.
  • Simplifies setup and maintenance of fluorescence compensation for the most common BD fluorochromes.
  • Increases compensation accuracy of BD FACSVerse™ flow cytometers.
  • Barcoding allows automatic entry of bead-lot-specific spillover factors.
  • Use this kit to set compensation for the BD Horizon™ V450 dye on BD flow cytometers with a violet laser.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Polystyrene beads, phosphate buffered saline with BSA and 0.1% sodium azide.