Learn More
BD Biosciences™ FC Bead 4c Plus Research Kit (RUO)
Click to view available options
:
To set compensation for tandem dyes PerCP-Cy™ 5.5, PE-Cy™7, APC-Cy7, and APC-H7 on BD flow cytometers with red and blue lasers
Description
- Designed for use on BD™ flow cytometers running BD FACSuite™ software.
- FC beads enable the software to automatically determine the spillover values for reference settings.
- Simplifies setup and maintenance of fluorescence compensation for the most common BD fluorochromes.
- Increases compensation accuracy of BD FACSVerse™ flow cytometers.
- Barcoding allows automatic entry of bead-lot-specific spillover factors.
- Use this kit to set compensation for tandem dyes PerCP-Cy™5.5, PE-Cy™7, APC-Cy7, and APC-H7 on BD flow cytometers with red and blue lasers.
Specifications
Specifications
| Content And Storage | FC beads (negative beads and positive capture beads), PerCP-Cy5.5-conjugated antibody, PE-Cy7-conjugated antibody, APC-Cy7-conjugated antibody, APC-H7-conjugated antibody, APC-H7-conjugated antibody, bead diluent, bead lot file card |
| Stability | The beads and reagents are stable until the expiration date shown on the bottle label when stored as directed. Do not use after the expiration date. |
| Product Type | Fluorescent Polystyrene Beads |
Safety and Handling
Product Identifier
- BD FACSUITE FC BEADS APC-H7 (RUO)
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- MIXTURE LIST-Contains : Sodium azide