missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ FC 11

Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
FC 11 is a highly potent focal adhesion kinase (FAK) PROTAC™ Degrader (DC50 values are 40 to 370 pM depending on cell line), which is composed of the FAK inhibitor PF 562217 joined by a linker to the cereblon-binding ligand Pomalidomide (Cat. No. 6302). The effects of FC 11 are reversible upon compound wash out. FC 11 also degrades autophosphorylated FAK (pFAKtyr397), displaying near complete degradation after 3 hours at 100 nM in TM3 cells.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 1-[2-[2-[2-[[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1 H-isoindol-4-yl]amino]ethoxy]ethoxy]ethyl]-N-[4-[[4-[[[2-[methyl(methylsulfonyl)amino]-3-pyridinyl]methyl]amino]-5-(trifluoromethyl)-2-pyrimidinyl]amino]phenyl]-1 H-1,2,3-triazole-4-carboxamid |
| CAS | 2271035-37-9 |
| Quantity | 2 mg |
| Target | FAK |
| Molecular Formula | C41H42F3N13O9S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction