missing translation for 'onlineSavingsMsg'
Learn More

FBXW8 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA544382

Code : NEW

Additional Details : Weight : 0.00010kg

Product Code. 15526482

  • £404.00 / 100µL
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Peptide sequence: MNQKLWEVYS GHPVQHISFS SHSLITANVP YQTVMRNADL DSFTTHRRHR Sequence homology: Cow: 83%; Dog: 92%; Guinea Pig: 83%; Horse: 92%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 92%

FBXW8 Polyclonal antibody specifically detects FBXW8 in Human samples. It is validated for Western Blot.
Specifications

Specifications

FBXW8
Polyclonal
Unconjugated
FBXW8
4930438M06Rik; F-box and WD repeat domain containing 8; F-box and WD-40 domain protein 8; F-box and WD-40 domain-containing protein 8; F-box only protein 29; F-box protein FBX29; F-box/WD repeat-containing protein 8; FBW6; FBW8; Fbx29; FBXO29; FBXW6; FBXW8; LOW QUALITY PROTEIN: F-box/WD repeat-containing protein 8; MGC33534
Rabbit
Affinity Chromatography
RUO
26259
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8N3Y1
FBXW8
Synthetic peptide directed towards the middle region of human FBXW8
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers