missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXW8 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94072-0.02ml
This item is not returnable.
View return policy
Description
FBXW8 Polyclonal antibody specifically detects FBXW8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| FBXW8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| F-box and WD repeat domain containing 8, F-box only protein 29F-box/WD repeat-containing protein 8, FBW8F-box and WD-40 domain-containing protein 8, FBX29MGC33534, FBXO29, FBXW6FBW6F-box and WD-40 domain protein 8 | |
| A synthetic peptide corresponding to a sequence within amino acids 500-598 of human FBXW8 (NP_699179.2). WDYRMNQKLWEVYSGHPVQHISFSSHSLITANVPYQTVMRNADLDSFTTHRRHRGLIRAYEFAVDQLAFQSPLPVCRSSCDAMATHYYDLALAFPYNHV | |
| 0.02 mL | |
| Cell Biology | |
| 26259 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction