missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXW4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10657-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a member of the F-box/WD-40 gene family, which recruit specific target proteins through their WD-40 protein-protein binding domains for ubiquitin mediated degradation. In mouse, a highly similar protein is thought to be responsible for maintaining the apical ectodermal ridge of developing limb buds; disruption of the mouse gene results in the absence of central digits, underdeveloped or absent metacarpal/metatarsal bones and syndactyly. This phenotype is remarkably similar to split hand-split foot malformation in humans, a clinically heterogeneous condition with a variety of modes of transmission. An autosomal recessive form has been mapped to the chromosomal region where this gene is located, and complex rearrangements involving duplications of this gene and others have been associated with the condition. A pseudogene of this locus has been mapped to one of the introns of the BCR gene on chromosome 22.Specifications
| FBXW4 | |
| Polyclonal | |
| Unconjugated | |
| FBXW4 | |
| DAC, Dactylin, FBW4, FBWD4, FBXW4, SHFM3, SHSF3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 30838, 309444, 6468 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P57775, Q9JMJ2 | |
| Fbxw4 | |
| FBXW4 Fusion Protein Ag1047 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Del en indholdskorrektion