missing translation for 'onlineSavingsMsg'
Learn More

FBXW11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16881214 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16881214 20 μL 20µL
16871214 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16881214 Supplier Proteintech Supplier No. 131491AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbws class and, in addition to an F-box, contains multiple WD40 repeats. This gene contains at least 14 exons, and its alternative splicing generates 3 transcript variants diverging at the presence/absence of two alternate exons.

Specifications

Antigen FBXW11
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FBXW11
Gene Accession No. Q5SRY7, Q9UKB1
Gene Alias BTRC2, BTRCP2, Fbw11, FBW1B, FBXW11, FBXW1B, Homologous to Slimb protein, Hos, KIAA0696
Gene Symbols Fbxw11
Host Species Rabbit
Immunogen FBXW11 Fusion Protein Ag3761
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103583, 23291, 303024
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.