missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11903-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. It contains a C-terminal amino acid sequence that bears a significant similarity with a portion of yeast Sec7p, a critical regulator of vesicular protein transport. This human protein may interact with ADP-ribosylation factor(s)(ARFs) and exhibit ARF-GEF (guanine nucleotide exchange factor) activity.Specifications
| FBXO8 | |
| Polyclonal | |
| Unconjugated | |
| FBXO8 | |
| DC10, F box only protein 8, F box protein 8, F box/SEC7 protein FBS, FBS, FBX8, FBXO8, UNQ1877/PRO4320 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26269, 306436, 50753 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NRD0, Q9QZN3 | |
| Fbxo8 | |
| FBXO8 Fusion Protein Ag2539 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction