Learn More
Invitrogen™ FBXO8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114038
Description
Immunogen sequence: EFFRHIHAPE ERGEYLETLI TKFSHRFCAC NPDLMRELGL SPDAVYVLCY For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. It contains a C-terminal amino acid sequence that bears a significant similarity with a portion of yeast Sec7p, a critical regulator of vesicular protein transport. This human protein may interact with ADP-ribosylation factor(s)(ARFs) and exhibit ARF-GEF (guanine nucleotide exchange factor) activity.
Specifications
| FBXO8 | |
| Polyclonal | |
| Unconjugated | |
| Fbxo8 | |
| DC10; F-box only protein 8; F-box protein 8; F-box protein Fbx8; F-box/SEC7 protein FBS; FBS; Fbx8; FBXO8; UNQ1877/PRO4320 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26269 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NRD0 | |
| Fbxo8 | |
| Synthetic peptide directed towards the middle region of human FBXO8. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.