missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO6 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98796UV
This item is not returnable.
View return policy
Description
FBXO6 Polyclonal antibody specifically detects FBXO6 in Human samples. It is validated for Western Blot
Specifications
| FBXO6 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 350 | |
| FBG2F-box/G-domain protein 2, F-box only protein 6, F-box protein 6, F-box protein Fbx6, F-box protein that recognizes sugar chains 2, Fbx6b, FBX6FBS2F-box protein FBG2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human FBXO6. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 26270 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction