missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10872-1-AP-150UL
This item is not returnable.
View return policy
Description
F-box proteins, critical components of the conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Novel human F-box proteins named FBG1, FBG2, FBG3, FBG4 and FBG5 constitute a subfamily of mammalian F-box proteins. FBG genes are expressed in a limited number of human tissues including kidney, liver, brain and muscle tissues.Specifications
| FBXO5 | |
| Polyclonal | |
| Unconjugated | |
| FBXO5 | |
| Early mitotic inhibitor 1, EMI1, F box only protein 5, F box protein 5, FBX5, FBXO5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26271, 292263, 67141 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7TSG3, Q9UKT4 | |
| Fbxo5 | |
| FBXO5 Fusion Protein Ag1296 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction