missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO48 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562040
This item is not returnable.
View return policy
Description
Immunogen sequence: SKRNNNLRVS HTEANSVDAE KEKNESQNNF FELLPAEITF KIFSQLDIRS LCRASL Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 55%.
FBXO48 gene ontology annotations related to this gene include SCF-dependent proteasomal ubiquitin-dependent protein catabolic process.
Specifications
| FBXO48 | |
| Polyclonal | |
| Unconjugated | |
| FBXO48 | |
| A630050E13Rik; F-box only protein 48; F-box protein 48; FBX48; Fbxo48; novel F-box domain containing protein; RGD1309073 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 554251 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5FWF7 | |
| FBXO48 | |
| Recombinant protein corresponding to Human FBXO48. Recombinant protein control fragment (Product #RP-99708). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction