missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO47 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548173
This item is not returnable.
View return policy
Description
FBXO47 Polyclonal Antibody for IHC (P), Western Blot
Probably recognizes and binds to some phosphorylated proteins and promotes their ubiquitination and degradation.
Specifications
| FBXO47 | |
| Polyclonal | |
| Unconjugated | |
| FBXO47 | |
| 2900052P03Rik; F-box only protein 47; F-box protein 47; Fbxo47; Gm1562 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 494188, 72973 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| A2A6H3, Q5MNV8 | |
| FBXO47 | |
| KLH conjugated synthetic peptide between 114-143 amino acids from the N-terminal region of human FBXO47. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction