missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO44 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582085
This item is not returnable.
View return policy
Description
Immunogen sequence: SLHRNLLHNP CAEEGFEFWS LDVNGGDEWK VEDLSRDQRK EFPNDQVRSQ ARLRVQVPAV RSAPVVRARA SGDLPARPGD HPAEERCQVE GGLPHILQLP Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 45%.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs family, similar to F-box only protein 2 and F-box only protein 6. Four alternatively spliced transcript variants encoding two distinct isoforms have been found for this gene.
Specifications
| FBXO44 | |
| Polyclonal | |
| Unconjugated | |
| FBXO44 | |
| 5730411K09; AV001623; DKFZp781J0852; F Box Gama 3; FBG3; F-box gene 3; F-box only protein 44; F-box only protein 6a; F-box protein 44; F-box protein FBX30; F-box/G-domain protein 3; FBX30; Fbx44; FBX6A; FBXO44; FBXO6A; MGC14140; RGD1562463; RP3-330O12.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 93611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H4M3 | |
| FBXO44 | |
| Recombinant protein corresponding to Human FBXO44. Recombinant protein control fragment (Product #RP-102200). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction