missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO44 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-51915
This item is not returnable.
View return policy
Description
FBXO44 Polyclonal specifically detects FBXO44 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications
| FBXO44 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FBG3 DKFZp781J0852, F-box gene 3, F-box only protein 44, F-box protein 44, F-box protein FBX30, F-box/G-domain protein 3, FBX30FBX6AFbx44, FBX44, Fbxo6a, MGC14140 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 93611 | |
| Human | |
| IgG |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
| 0.5 mg/mL | |
| Flow Cytometry 10 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Peptide ELISA Detection limit 1:8000 | |
| NP_904319.1 | |
| FBXO44 | |
| Peptide with sequence C-AALTPPEPPSAEP corresponding to C-Terminus according to NP_904319.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize isoform 2 (NP_904319.1 and NP_9043201 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction