missing translation for 'onlineSavingsMsg'
Learn More

FBXO44 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-467MFV450

Product Code. 18721223

  • £393.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FBXO44 Polyclonal antibody specifically detects FBXO44 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FBXO44
Polyclonal
Western Blot
FBG3 DKFZp781J0852, F-box gene 3, F-box only protein 44, F-box protein 44, F-box protein FBX30, F-box/G-domain protein 3, FBX30FBX6AFbx44, FBX44, Fbxo6a, MGC14140
A synthetic peptide containing residues 103-119 EWKVEDLSRDQRKEFPN) of the human FBG3 protein.
0.1 mL
Primary
Human
Purified
Western Blot
mFluor Violet 450 SE
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
93611
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.