missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO42 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA555224
This item is not returnable.
View return policy
Description
Immunogen sequence: HSLPPIARRL GHHPPQSLNV GKPLYQSMNC KPMQMYVLDI KDTKEKGRVK WKVFNSSSVV GPPETSLHTV VQGRGELIIF GGLMDKKQNV KY Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
Members of the F-box protein family, such as FBXO42, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
Specifications
| FBXO42 | |
| Polyclonal | |
| Unconjugated | |
| FBXO42 | |
| 6720460I06Rik; F-box only protein 42; F-box protein 42; Fbx42; Fbxo42; JFK; just one F-box and Kelch domain-containing protein; KIAA1332; LOC527807 protein; mKIAA1332 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54455 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P3S6 | |
| FBXO42 | |
| Recombinant protein corresponding to Human FBXO42 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction