missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565932
This item is not returnable.
View return policy
Description
Immunogen sequence: QVLELDHVSEI TQEVAAEVCR EGLKGLEMLV LTATPVTPKA LLHFNSICRN LKSIVVQIGI ADYFKEPSSP EAQKLFEDMV TKLQALRRRP GFSKILH Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Substrate-recognition component of the SCF (SKP1-CUL1-F- box protein)-type E3 ubiquitin ligase complex.
Specifications
| FBXO41 | |
| Polyclonal | |
| Unconjugated | |
| FBXO41 | |
| 9630017H13; D6Ertd538e; F-box only protein 41; F-box protein 41; FBX41; FBXO41; Kiaa1940; RGD1566130 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 150726 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TF61 | |
| FBXO41 | |
| Recombinant Human FBXO41. Recombinant protein control fragment (Product #RP-106613). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion