missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FBXO41 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA548401

Product Code. 15682695

  • £443.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FBXO41 Polyclonal Antibody for Western Blot

Substrate-recognition component of the SCF (SKP1-CUL1-F- box protein)-type E3 ubiquitin ligase complex.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FBXO41
Polyclonal
Unconjugated
FBXO41
9630017H13; D6Ertd538e; F-box only protein 41; F-box protein 41; FBX41; FBXO41; Kiaa1940; RGD1566130
Rabbit
Antigen affinity chromatography, Protein A
RUO
330369
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q6NS60
FBXO41
KLH conjugated synthetic peptide between 235-263 amino acids from the N-terminal region of human FBX41.
400 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.