missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566167
This item is not returnable.
View return policy
Description
Immunogen sequence: ARREVFESTSF QGKEQAAGPS PAAPHLLHHH HHHAPLAHFP GDLVPASLPC EELAEPGLVP AAAARYALRE IEIPLGELFA RKSVAS Highest antigen sequence identity to the following orthologs - mouse 94%, rat 95%.
Substrate-recognition component of the SCF (SKP1-CUL1-F- box protein)-type E3 ubiquitin ligase complex.
Specifications
| FBXO41 | |
| Polyclonal | |
| Unconjugated | |
| FBXO41 | |
| 9630017H13; D6Ertd538e; F-box only protein 41; F-box protein 41; FBX41; FBXO41; Kiaa1940; RGD1566130 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 150726 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TF61 | |
| FBXO41 | |
| Recombinant Human FBXO41. Recombinant protein control fragment (Product #RP-106851). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction