missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FBXO36 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA565825

Product Code. 15998554

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RLSDDLLLTII SYLDLEDIAR LCQTSHRFAK LCMSDKLWEQ IVQSTCDTIT PDVRALAEDT GWRQLFFTNK LQLQRQLRKR KQ Highest antigen sequence identity to the following orthologs - mouse 76%, rat 80%.

Members of the F-box protein family, such as FBXO36, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FBXO36
Polyclonal
Unconjugated
FBXO36
0610008D19Rik; 1110020F21Rik; 2410002G19Rik; D1Ertd757e; F-box only protein 36; F-box protein 36; Fbx36; Fbxo36
Rabbit
Antigen affinity chromatography
RUO
130888
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8NEA4
FBXO36
Recombinant Human FBXO36. Recombinant protein control fragment (Product #RP-104979).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.