missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO31 Antibody (OTI1H1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72208CL1
This item is not returnable.
View return policy
Description
FBXO31 Monoclonal antibody specifically detects FBXO31 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| FBXO31 | |
| Monoclonal | |
| CoraFluor 1 | |
| DKFZp434B027, F-box only protein 31, F-box protein 31, FBX14DKFZp434J1815, Fbx31, FBXO14FLJ22477, MGC15419, MGC9527, pp2386, putative breast cancer tumor-suppressor, SCF ubiquitin ligase specificity factor | |
| Full length human recombinant protein of human FBXO31(NP_079011) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI1H1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 79791 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction