missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO31 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-19088
This item is not returnable.
View return policy
Description
FBXO31 Polyclonal specifically detects FBXO31 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation (Negative).
Specifications
| FBXO31 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DKFZp434B027, F-box only protein 31, F-box protein 31, FBX14DKFZp434J1815, Fbx31, FBXO14FLJ22477, MGC15419, MGC9527, pp2386, putative breast cancer tumor-suppressor, SCF ubiquitin ligase specificity factor | |
| Rabbit | |
| 60.7 kDa | |
| 100 μL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, African elephant, Naked mole rat and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:5000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Q5XUX0 | |
| FBXO31 | |
| The immunogen recognized by this antibody maps to a region between residue 489 and 539 of human F-box protein 31 using the numbering given in entry NP_079011.3 (GeneID 79791). | |
| Affinity Purified | |
| RUO | |
| 79791 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction