missing translation for 'onlineSavingsMsg'
Learn More

FBXO31 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-19088F

Product Code. 18515711

  • £529.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FBXO31 Polyclonal antibody specifically detects FBXO31 in Human, Mouse samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FBXO31
Polyclonal
Immunocytochemistry/Immunofluorescence
DKFZp434B027, F-box only protein 31, F-box protein 31, FBX14DKFZp434J1815, Fbx31, FBXO14FLJ22477, MGC15419, MGC9527, pp2386, putative breast cancer tumor-suppressor, SCF ubiquitin ligase specificity factor
The immunogen recognized by this antibody maps to a region between residue 489 and 539 of human F-box protein 31 using the numbering given in entry NP_079011.3 (GeneID 79791).
0.1 mL
Primary
Human, Mouse
Purified
Immunofluorescence
FITC
PBS
Rabbit
Immunogen affinity purified
RUO
79791
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.