missing translation for 'onlineSavingsMsg'
Learn More

FBXO31 Antibody (1C5), Novus Biologicals™

Product Code. 18407889 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407889 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407889 Supplier Novus Biologicals Supplier No. H00079791M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FBXO31 Monoclonal antibody specifically detects FBXO31 in Human samples. It is validated for ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO31
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1C5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_079011
Gene Alias DKFZp434B027, F-box only protein 31, F-box protein 31, FBX14DKFZp434J1815, Fbx31, FBXO14FLJ22477, MGC15419, MGC9527, pp2386, putative breast cancer tumor-suppressor, SCF ubiquitin ligase specificity factor
Host Species Mouse
Immunogen FBXO31 (NP_079011.3, 269 a.a. ∽ 367 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GQGQPFVLPVGVSSRNEDYPRTCRMCFYGTGLIAGHGFTSPERTPGVFILFDEDRFGFVWLELKSFSLYSRVQATFRNADAPSPQAFDEMLKNIQSLTS
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79791
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.