missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17803-1-AP-20UL
This item is not returnable.
View return policy
Description
FBXO3 encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs.Specifications
| FBXO3 | |
| Polyclonal | |
| Unconjugated | |
| FBXO3 | |
| F box only protein 3, F box protein 3, FBA, FBX3, FBXO3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26273, 57443, 690634 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D4ABP9, Q9DC63, Q9UK99 | |
| Fbxo3 | |
| FBXO3 Fusion Protein Ag12071 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction