Learn More
Invitrogen™ FBXO3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116640
Description
Positive Samples: Mouse liver, Mouse lung, Mouse eye Immunogen sequence: MAAMETETAP LTLESLPTDP LLLILSFLDY RDLINCCYVS RRLSQLSSHD PLWRRHCKKY WLISEEEKTQ KNQCWKSLFI DTYSDVGRYI DHYAAIKKAW DDLKKYLEPR CPRMVLSLKE GAREEDLDAV EAQIGCKLPD DYRCSYRIHN GQKLVVPGLL GSMALSNHYR SEDLLDVDTA AGGFQQRQGL KYCLPLTFCI HTGLSQYIAV EAAEGRNKNE VFYQCPDQMA RNPAAIDMFI.
FBXO3 encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs.
Specifications
| FBXO3 | |
| Polyclonal | |
| Unconjugated | |
| FBXO3 | |
| 1200002G09Rik; 1700026K02Rik; AI046358; DKFZp564B092; FBA; F-box only protein 3; F-box protein 3; F-box protein FBA; F-box protein FBX3; FBX3; FBXO3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57443 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.14 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9DC63 | |
| FBXO3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human FBXO3 (NP_0363072). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.