missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO28 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112110
This item is not returnable.
View return policy
Description
FBXO28 Polyclonal Antibody for Western Blot, ICC/IF
Members of the F-box protein family, such as FBXO28, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.
Specifications
| FBXO28 | |
| Polyclonal | |
| Unconjugated | |
| FBXO28 | |
| 4833428J17Rik; 5730505P19Rik; CENP-30; centromere protein 30; D1Ertd578e; F-box only protein 28; F-box protein 28; Fbx28; FBXO28; KIAA0483; mKIAA0483 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 23219 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9NVF7 | |
| FBXO28 | |
| A synthetic peptide corresponding to the C-terminus of the Human FBXO28. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction