Learn More
Invitrogen™ FBXO25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113762
Description
Immunogen sequence: LGEAFNRLDF SSAIQDIRRF NYVVKLLQLI AKSQLTSLSG VAQKNYFNIL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 93%; Zebrafish: 93%.
FBXO25 is a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. FBXO25 belongs to the Fbxs class.
Specifications
| FBXO25 | |
| Polyclonal | |
| Unconjugated | |
| FBXO25 | |
| 9130015I06Rik; AI649137; F-box only protein 25; F-box protein 25; F-box protein Fbx25; FBX25; FBXO25; MGC20256; MGC51975; Unknown (protein for MGC:137945); zgc:100907 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26260 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TCJ0 | |
| FBXO25 | |
| Synthetic peptide directed towards the N terminal region of human FBXO25 (aa 103-152). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.