missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FBXO24 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA562275

Product Code. 15730072

  • £437.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: IYSLVVNETQ LDQPRSYTVQ LALRKVSHYL PHLRVACMTS NQSSTLYVTD QGGVYFEVHT PGVYRDLFGT LQA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FBXO24
Polyclonal
Unconjugated
FBXO24
4933422D21Rik; DKFZp434I1122; F-box only protein 24; F-box protein 24; F-box protein Fbx24; Fbx24; FBXO24; LOW QUALITY PROTEIN: F-box only protein 24
Rabbit
Antigen affinity chromatography
RUO
26261
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O75426
FBXO24
Recombinant protein corresponding to Human FBXO24. Recombinant protein control fragment (Product #RP-101172).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.