missing translation for 'onlineSavingsMsg'
Learn More

FBXO22 Antibody (OTI1H1) [DyLight 488], Novus Biologicals™

Product Code. 30688956 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688956 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688956 Supplier Novus Biologicals Supplier No. NBP345022G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FBXO22 Monoclonal antibody specifically detects FBXO22 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO22
Applications Western Blot
Classification Monoclonal
Clone OTI1H1
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias F-box only protein 22, F-box protein 22, F-box protein FBX22p44, FBX22FLJ13986, FIST domain containing 1, FISTC1, MGC31799
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-220 of human FBXO22 (NP_671717) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 26263
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.