missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO22 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16447
This item is not returnable.
View return policy
Description
FBXO22 Polyclonal specifically detects FBXO22 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FBXO22 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| F-box only protein 22, F-box protein 22, F-box protein FBX22p44, FBX22FLJ13986, FIST domain containing 1, FISTC1, MGC31799 | |
| Rabbit | |
| 45 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 0.29mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q8NEZ5 | |
| FBXO22 | |
| Recombinant protein encompassing a sequence within the center region of human FBXO22. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 26263 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction